missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylnaphthalene (stabilized with TBC) 95.0+%, TCI America™

Catalog No. V00405G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V00405G Supplier TCI America Supplier No. V00405G

Chemical Identifiers

CAS 826-74-4
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00075766
InChI Key IGGDKDTUCAWDAN-UHFFFAOYSA-N
Synonym 1-vinylnaphthalene, vinylnaphthalene, 1-vinyl naphthalene, unii-pf20cj2k0n, pf20cj2k0n, 1-vinylphthalene, 1-vinyl-naphthalene, naphthalene, ethenyl, naphthalen-1-yl ethene, naphthalene, 1-ethenyl
PubChem CID 70004
ChEBI CHEBI:51327
IUPAC Name 1-ethenylnaphthalene
SMILES C=CC1=C2C=CC=CC2=CC=C1

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
Formula Weight 154.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Vinylnaphthalene (stabilized with TBC)
TSCA No
Recommended Storage Freezer (-20 °C)