missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylnaphthalene, 95%, stab. with 4-tert-butylcatechol

Catalog No. p-7033865
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5104303 1 g
AAH5104306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5104303 Supplier Thermo Scientific Chemicals Supplier No. H5104303

CAS: 826-74-4 | C12H10 | 154.21 g/mol

1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.

Solubility
Soluble in ethyl acetate, hexane and methanol.

Notes
Moisture sensitive. Incompatible with oxidizing agents, heat, light, ultraviolet radiation and free radical initiators.

Chemical Identifiers

CAS 826-74-4
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00075766
InChI Key IGGDKDTUCAWDAN-UHFFFAOYSA-N
Synonym 1-vinylnaphthalene, vinylnaphthalene, 1-vinyl naphthalene, unii-pf20cj2k0n, pf20cj2k0n, 1-vinylphthalene, 1-vinyl-naphthalene, naphthalene, ethenyl, naphthalen-1-yl ethene, naphthalene, 1-ethenyl
PubChem CID 70004
ChEBI CHEBI:51327
IUPAC Name 1-ethenylnaphthalene
SMILES C=CC1=C2C=CC=CC2=CC=C1

Specifications

Density 1.04
Boiling Point 135°C to 138°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in ethyl acetate,hexane and methanol.
Formula Weight 154.21
Percent Purity 95%
Chemical Name or Material 1-Vinylnaphthalene, stabilized with 4-tert-butylcatechol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-560-5
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only