missing translation for 'onlineSavingsMsg'
Learn More

1-Undecyne 96.0+%, TCI America™

Catalog No. U00335ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00335ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00335ML Supplier TCI America Supplier No. U00335ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2243-98-3
Molecular Formula C11H20
Molecular Weight (g/mol) 152.281
MDL Number MFCD00015063
InChI Key YVSFLVNWJIEJRV-UHFFFAOYSA-N
Synonym 1-undecyne, rutylidene, unii-y07u3q24us, undecyne, acmc-1cr7r, 1-undecyne 10g
PubChem CID 75249
ChEBI CHEBI:87545
IUPAC Name undec-1-yne
SMILES CCCCCCCCCC#C

Specifications

Color Colorless
Boiling Point 195°C
Quantity 5 mL
Formula Weight 152.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Undecyne