missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Undecyne, 97%
Description
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.
Solubility
Miscible with acetone, ether, ethanol and benzene. Slightly miscible with water.
Notes
Incompatible with oxidizing agents.
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.
Solubility
Miscible with acetone, ether, ethanol and benzene. Slightly miscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -25°C |
| Density | 0.77 |
| Boiling Point | 196°C |
| Flash Point | 65°C (149°F) |
| Odor | Characteristic |
| Refractive Index | 1.431 |
| Linear Formula | CH3(CH2)8C≡CH |
| Quantity | 10 g |
| Beilstein | 1740043 |
| Synonym | 1-undecyne, rutylidene, unii-y07u3q24us, undecyne, acmc-1cr7r, 1-undecyne 10g |
| Show More |
Safety and Handling
| Hazard Category | H227 |
| Precautionary Statement | P210-P280-P370+P378q-P501c |
| EINECSNumber | 218-825-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only