missing translation for 'onlineSavingsMsg'
Learn More

1-Undecyne, 97%

Catalog No. AAL0288318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0288318 50 g
AAL0288309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0288318 Supplier Thermo Scientific Chemicals Supplier No. L0288318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2243-98-3 | C11H20 | 152.281 g/mol

1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Undecyne is used as a reagent organic synthesis. Further, it is used in the preparation of an antimalarial and antitrypanosomal agent.

Solubility
Miscible with acetone, ether, ethanol and benzene. Slightly miscible with water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2243-98-3
Molecular Formula C11H20
MDL Number MFCD00015063
InChI Key YVSFLVNWJIEJRV-UHFFFAOYSA-N
PubChem CID 75249
ChEBI CHEBI:87545
IUPAC Name undec-1-yne
SMILES CCCCCCCCCC#C

Specifications

Melting Point -25°C
Density 0.77
Boiling Point 196°C
Flash Point 65°C (149°F)
Odor Characteristic
Refractive Index 1.431
Linear Formula CH3(CH2)8C≡CH
Quantity 50 g
Beilstein 1740043
Synonym 1-undecyne, rutylidene, unii-y07u3q24us, undecyne, acmc-1cr7r, 1-undecyne 10g
Solubility Information Miscible with acetone,ether,ethanol and benzene. Slightly miscible with water.
Molecular Weight (g/mol) 152.281
Formula Weight 152.28
Percent Purity 97%
Chemical Name or Material 1-Undecyne
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 218-825-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only