missing translation for 'onlineSavingsMsg'
Learn More

1-Undecanethiol 96.0+%, TCI America™

Catalog No. U00665ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00665ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00665ML Supplier TCI America Supplier No. U00665ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used in materials science research for applications such as self-assembly and as a capping agent.

Chemical Identifiers

CAS 5332-52-5
Molecular Formula C11H24S
Molecular Weight (g/mol) 188.37
MDL Number MFCD00022097
InChI Key CCIDWXHLGNEQSL-UHFFFAOYSA-N
Synonym Undecyl Mercaptan
PubChem CID 62570
IUPAC Name undecane-1-thiol
SMILES CCCCCCCCCCCS

Specifications

Melting Point -3°C
Color Colorless
Boiling Point 104°C
Quantity 5 mL
Formula Weight 188.37
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Undecanethiol
EINECSNumber (2)-0464
TSCA No