missing translation for 'onlineSavingsMsg'
Learn More

1-Trityl-L-histidine, 98%

CAS: 35146-32-8 | C25H23N3O2 | 397.478 g/mol

$100.45 - $347.77

Chemical Identifiers

CAS 35146-32-8
Molecular Formula C25H23N3O2
Molecular Weight (g/mol) 397.478
MDL Number MFCD00153444
InChI Key BSZQZNOAYQCQFZ-QHCPKHFHSA-N
Synonym h-his trt-oh, 1-triphenylmethyl-l-histidine, l-histidine, 1-triphenylmethyl, s-2-amino-3-1-trityl-1h-imidazol-4-yl propanoic acid, 2s-2-amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, 1-trityl-l-histidine, pubchem18948, n-im-trityl-l-histidine, n im-trityl-l-histidine
PubChem CID 7408287
IUPAC Name (2S)-2-amino-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6277903
View Documents
Thermo Scientific Chemicals
H6277903
1 g
Each for $100.45
Only null left
 
AAH6277906
View Documents
Thermo Scientific Chemicals
H6277906
5 g
Each for $347.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35146-32-8
397.478
BSZQZNOAYQCQFZ-QHCPKHFHSA-N
7408287
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)N
C25H23N3O2
MFCD00153444
h-his trt-oh, 1-triphenylmethyl-l-histidine, l-histidine, 1-triphenylmethyl, s-2-amino-3-1-trityl-1h-imidazol-4-yl propanoic acid, 2s-2-amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, 1-trityl-l-histidine, pubchem18948, n-im-trityl-l-histidine, n im-trityl-l-histidine
(2S)-2-amino-3-(1-tritylimidazol-4-yl)propanoic acid

Specifications

210°C (decomposition)
C25H23N3O2
767244
BSZQZNOAYQCQFZ-QHCPKHFHSA-N
(2S)-2-amino-3-(1-tritylimidazol-4-yl)propanoic acid
397.478
397.47
Solid
35146-32-8
MFCD00153444
h-his trt-oh, 1-triphenylmethyl-l-histidine, l-histidine, 1-triphenylmethyl, s-2-amino-3-1-trityl-1h-imidazol-4-yl propanoic acid, 2s-2-amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, 1-trityl-l-histidine, pubchem18948, n-im-trityl-l-histidine, n im-trityl-l-histidine
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)N
1 g
7408287
98%
1-Trityl-L-histidine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only