missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsilyloxy)cyclohexene 98.0+%, TCI America™

Catalog No. T155610ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T155610ML 10 mL
T155625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T155610ML Supplier TCI America Supplier No. T155610ML

Chemical Identifiers

CAS 6651-36-1
Molecular Formula C9H18OSi
Molecular Weight (g/mol) 170.33
MDL Number MFCD00001541
InChI Key SBEMOANGDSSPJY-UHFFFAOYSA-N
Synonym 1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane
PubChem CID 81161
IUPAC Name (cyclohex-1-en-1-yloxy)trimethylsilane
SMILES C[Si](C)(C)OC1=CCCCC1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 10 mL
UN Number 1993
Formula Weight 170.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Trimethylsilyloxy)cyclohexene