missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsilyl)imidazole, Approx. 97%, Spectrum™ Chemical

Catalog No. p-7157858
Change view
Click to view available options
Quantity:
25 mL
100 mL
40 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600011 25 mL Amber Glass Bottle
18600012 100 mL Amber Glass Bottle
18600013 40 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600011 Supplier Spectrum Chemical Mfg Cor Supplier No. T215725ML
Only null left

T2157, 18156-74-6, C6H12N2Si

Spectrum™ Chemical 1-(Trimethylsilyl)imidazole

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
IUPAC Name 1-(trimethylsilyl)-1H-imidazole
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 mL
UN Number UN1993
Percent Purity ∼97%
Grade Reagent