missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Trimethylsilyl)imidazole, Approx. 97%, Spectrum™ Chemical
Description
Spectrum™ Chemical 1-(Trimethylsilyl)imidazole
Specifications
Specifications
| CAS Max % | 100% |
| Boiling Point | 93°C to 94°C/199.4°F to 201.2°F |
| Packaging | Amber Glass Bottle |
| Quantity | 40 kg |
| UN Number | UN1993 |
| Percent Purity | ∼97% |
| Grade | Ungraded |
| Physical Form | Liquid |
| Chemical Name or Material | 1-(Trimethylsilyl)imidazole |