missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsilyl)imidazole, 97%

CAS: 18156-74-6 | C6H12N2Si | 140.26 g/mol

Supplier:  Thermo Scientific Chemicals A1251230

 View more versions of this product

Catalog No. AAA1251230


Only null left
Explore available promotions

Description

Description

1-(Trimethylsilyl)imidazole acts as a silylating agent for alcohols, carbohydrates and 1,3-dicarbonyl compounds. It is a derivatization reagent used for selective silylation of hydroxyl groups and carboxylic acid without affecting the amino group. It employed as an intermediate for the synthesis of imidazole derivatives. It plays an essential role to synthesize polysubstituted chiral spirotetrahydropyrans. It is also used in the quantification of pintol after derivatization by gas chromatography/ mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trimethylsilyl)imidazole acts as a silylating agent for alcohols, carbohydrates and 1,3-dicarbonyl compounds. It is a derivatization reagent used for selective silylation of hydroxyl groups and carboxylic acid without affecting the amino group. It employed as an intermediate for the synthesis of imidazole derivatives. It plays an essential role to synthesize polysubstituted chiral spirotetrahydropyrans. It is also used in the quantification of pintol after derivatization by gas chromatography/ mass spectrometry.

Solubility
Miscible with most common organic solvents. It decomposes in water.

Notes
Moisture and light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

1-(Trimethylsilyl)imidazole
-42°C
212°C
Weak
1.4765
250 g
606148
n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
YKFRUJSEPGHZFJ-UHFFFAOYSA-N
imidazol-1-yl(trimethyl)silane
28925
140.26
18156-74-6
0.956
18°C (64°F)
C6H12N2Si
MFCD00005280
UN1993
Moisture sensitive
Miscible with most common organic solvents. It decomposes in water.
C[Si](C)(C)N1C=CN=C1
140.26
CHEBI:85063
97%

Chemical Identifiers

18156-74-6
140.26
YKFRUJSEPGHZFJ-UHFFFAOYSA-N
28925
imidazol-1-yl(trimethyl)silane
C6H12N2Si
MFCD00005280
n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
CHEBI:85063
C[Si](C)(C)N1C=CN=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 242-040-3

RTECSNumber : NI8700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only