missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsilyl)imidazole, 97%

Catalog No. AAA1251206
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1251206 5 g
AAA1251218 50 g
AAA1251222 100 g
AAA1251230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1251206 Supplier Thermo Scientific Chemicals Supplier No. A1251206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18156-74-6 | C6H12N2Si | 140.26 g/mol

1-(Trimethylsilyl)imidazole acts as a silylating agent for alcohols, carbohydrates and 1,3-dicarbonyl compounds. It is a derivatization reagent used for selective silylation of hydroxyl groups and carboxylic acid without affecting the amino group. It employed as an intermediate for the synthesis of imidazole derivatives. It plays an essential role to synthesize polysubstituted chiral spirotetrahydropyrans. It is also used in the quantification of pintol after derivatization by gas chromatography/ mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trimethylsilyl)imidazole acts as a silylating agent for alcohols, carbohydrates and 1,3-dicarbonyl compounds. It is a derivatization reagent used for selective silylation of hydroxyl groups and carboxylic acid without affecting the amino group. It employed as an intermediate for the synthesis of imidazole derivatives. It plays an essential role to synthesize polysubstituted chiral spirotetrahydropyrans. It is also used in the quantification of pintol after derivatization by gas chromatography/ mass spectrometry.

Solubility
Miscible with most common organic solvents. It decomposes in water.

Notes
Moisture and light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name imidazol-1-yl(trimethyl)silane
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

Melting Point -42°C
Density 0.956
Boiling Point 212°C
Flash Point 18°C (64°F)
Odor Weak
Refractive Index 1.4765
Quantity 5 g
UN Number UN1993
Beilstein 606148
Sensitivity Moisture sensitive
Solubility Information Miscible with most common organic solvents. It decomposes in water.
Formula Weight 140.26
Percent Purity 97%
Chemical Name or Material 1-(Trimethylsilyl)imidazole
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 242-040-3
RTECSNumber NI8700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only