missing translation for 'onlineSavingsMsg'
Learn More

1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98.0+%, TCI America™

Catalog No. p-7137671
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32721G 1 g
T32725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32721G Supplier TCI America Supplier No. T32721G
Only null left

Chemical Identifiers

CAS 181934-29-2
Molecular Formula C12H13IO2Si
Molecular Weight (g/mol) 344.223
InChI Key KUCCHAADRXRBFZ-UHFFFAOYSA-N
Synonym TMS-EBX
PubChem CID 10641155
IUPAC Name 1-(2-trimethylsilylethynyl)-1$l^{3},2-benziodoxol-3-one
SMILES C[Si](C)(C)C#CI1C2=CC=CC=C2C(=O)O1

Specifications

Melting Point 140°C
Quantity 1 g
Formula Weight 344.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one
TSCA No
Recommended Storage Refrigerator