missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 140°C |
| Quantity | 1 g |
| Formula Weight | 344.22 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |