Learn More
1-(Trimethylsiloxy)cyclohexene, 98%
CAS: 6651-36-1 | C9H18OSi | 170.33 g/mol
Supplier: Thermo Scientific Chemicals A1609918
Description
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
Specifications
| 165°C | |
| 6651-36-1 | |
| C9H18OSi | |
| UN1993 | |
| 42°C (107°F) | |
| Not miscible or difficult to mix. | |
| C[Si](C)(C)OC1=CCCCC1 | |
| 170.33 | |
| 170.33 | |
| 1.447 | |
| 0.883 |
| 1-(Trimethylsiloxy)cyclohexene | |
| 50 g | |
| MFCD00001541 | |
| 1859394 | |
| 1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane | |
| SBEMOANGDSSPJY-UHFFFAOYSA-N | |
| cyclohexen-1-yloxy(trimethyl)silane | |
| 81161 | |
| 98% | |
| Moisture sensitive |
Chemical Identifiers
| 6651-36-1 | |
| 170.33 | |
| SBEMOANGDSSPJY-UHFFFAOYSA-N | |
| 81161 | |
| C[Si](C)(C)OC1=CCCCC1 |
| C9H18OSi | |
| MFCD00001541 | |
| 1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane | |
| cyclohexen-1-yloxy(trimethyl)silane |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 229-675-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only