missing translation for 'onlineSavingsMsg'
Learn More

1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98.0+%, TCI America™

Catalog No. p-7137541
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3039200MG 200 mg
T30391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3039200MG Supplier TCI America Supplier No. T3039200MG
Only null left

Chemical Identifiers

CAS 181934-30-5
Molecular Formula C18H25IO2Si
Molecular Weight (g/mol) 428.385
MDL Number MFCD18632570
InChI Key NTHGHMCOPNSZIR-UHFFFAOYSA-N
Synonym TIPS-EBX
PubChem CID 10693803
IUPAC Name 1-[2-tri(propan-2-yl)silylethynyl]-1$l^{3},2-benziodoxol-3-one
SMILES CC(C)[Si](C#CI1C2=CC=CC=C2C(=O)O1)(C(C)C)C(C)C

Specifications

Melting Point 170°C
Color White
Quantity 200 mg
Formula Weight 428.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one