missing translation for 'onlineSavingsMsg'
Learn More

1-Trifluoromethylcyclopropane-1-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAL1795703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1795703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1795703 Supplier Thermo Scientific Chemicals Supplier No. L1795703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 277756-46-4 | Molecular Formula: C5H5F3O2 | Molecular Weight: 154.088 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 277756-46-4
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.088
MDL Number MFCD03093070
InChI Key SKCBKBCACWDALV-UHFFFAOYSA-N
Synonym 1-trifluoromethyl cyclopropane-1-carboxylic acid, 1-trifluoromethyl cyclopropanecarboxylic acid, 1-trifluoromethylcyclopropanecarboxylic acid, 1-trifluoromethyl cyclopropane-1-carboxylicacid, 1-trifluoromethyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-trifluoromethyl, 1-trifluoromethyl-cyclopropanecarboxylic acid, pubchem20086, acmc-20aa6t, 1-trifluoromethylcyclopropanecarboxylicacid
PubChem CID 2778306
IUPAC Name 1-(trifluoromethyl)cyclopropane-1-carboxylic acid
SMILES C1CC1(C(=O)O)C(F)(F)F

Specifications

Melting Point 87°C to 89°C
Boiling Point 182°C to 184°C
Quantity 1 g
UN Number UN3261
Beilstein 8543283
Formula Weight 154.09
Percent Purity 97%
Chemical Name or Material 1-Trifluoromethylcyclopropane-1-carboxylic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only