Learn More
1-(Trifluoromethyl)cyclopentanecarboxylic acid, 97%
CAS: 277756-44-2 | C7H9F3O2 | 182.142 g/mol
$301.86 - $315.99
Chemical Identifiers
| CAS | 277756-44-2 |
|---|---|
| Molecular Formula | C7H9F3O2 |
| Molecular Weight (g/mol) | 182.142 |
| MDL Number | MFCD08445821 |
| InChI Key | DGQRYPPBAJNZFZ-UHFFFAOYSA-N |
| Synonym | 1-trifluoromethyl cyclopentane-1-carboxylic acid, 1-trifluoromethyl cyclopentanecarboxylic acid, acmc-1cp3t, 1-trifluoromethyl-cyclopentanecarboxylic acid, 1-trifluoromethylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-trifluoromethyl |
| PubChem CID | 15507525 |
| IUPAC Name | 1-(trifluoromethyl)cyclopentane-1-carboxylic acid |
| SMILES | C1CCC(C1)(C(=O)O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH26055MD
|
Thermo Scientific Chemicals
H26055MD |
250 mg |
Each for $301.86
|
|
|||||
|
AAH2605503
|
Thermo Scientific Chemicals
H2605503 |
1 g |
Each for $315.99
|
|
|||||
Description
1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Chemical Identifiers
| 277756-44-2 | |
| 182.142 | |
| DGQRYPPBAJNZFZ-UHFFFAOYSA-N | |
| 15507525 | |
| C1CCC(C1)(C(=O)O)C(F)(F)F |
| C7H9F3O2 | |
| MFCD08445821 | |
| 1-trifluoromethyl cyclopentane-1-carboxylic acid, 1-trifluoromethyl cyclopentanecarboxylic acid, acmc-1cp3t, 1-trifluoromethyl-cyclopentanecarboxylic acid, 1-trifluoromethylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-trifluoromethyl | |
| 1-(trifluoromethyl)cyclopentane-1-carboxylic acid |
Specifications
| 277756-44-2 | |
| C7H9F3O2 | |
| 250 mg | |
| Insoluble in water. | |
| C1CCC(C1)(C(=O)O)C(F)(F)F | |
| 182.142 | |
| 182.15 | |
| 1-(Trifluoromethyl)cyclopentanecarboxylic acid |
| 36°C to 38°C | |
| MFCD08445821 | |
| 1-trifluoromethyl cyclopentane-1-carboxylic acid, 1-trifluoromethyl cyclopentanecarboxylic acid, acmc-1cp3t, 1-trifluoromethyl-cyclopentanecarboxylic acid, 1-trifluoromethylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-trifluoromethyl | |
| DGQRYPPBAJNZFZ-UHFFFAOYSA-N | |
| 1-(trifluoromethyl)cyclopentane-1-carboxylic acid | |
| 15507525 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only