missing translation for 'onlineSavingsMsg'
Learn More

1-Trifluoromethyl-1,2-benziodoxol-3(1H)-one (contains 60% Diatomaceous earth), TCI America™

Catalog No. T30141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30141G Supplier TCI America Supplier No. T30141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 887144-94-7
Molecular Formula C8H4F3IO2
Molecular Weight (g/mol) 316.018
MDL Number MFCD18800706
InChI Key XHEOXSQMBWJOKP-UHFFFAOYSA-N
Synonym Togni Reagent II
PubChem CID 24850981
IUPAC Name 1-(trifluoromethyl)-1$l^{3},2-benziodoxol-3-one
SMILES C1=CC=C2C(=C1)C(=O)OI2C(F)(F)F

Specifications

Melting Point 158°C
Color White
Quantity 1 g
UN Number 1325
Formula Weight 316.02
Physical Form Crystalline Powder
Chemical Name or Material 1-Trifluoromethyl-1,2-benziodoxol-3(1H)-one (contains 60% Diatomaceous earth)
TSCA No
Recommended Storage Refrigerator