missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Trifluoroacetyl)imidazole 98.0+%, TCI America™
Supplier: TCI America T067025G
Specifications
| 1-(Trifluoroacetyl)imidazole | |
| 83°C | |
| MFCD00014501 | |
| 1993 | |
| SINBGNJPYWNUQI-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-imidazol-1-ylethanone | |
| 73767 | |
| ≥98.0% (T) |
| 1546-79-8 | |
| C5H3F3N2O | |
| 25 g | |
| 1-trifluoroacetyl imidazole, n-trifluoroacetylimidazole, 1-trifluoroacetyl-1h-imidazole, 1h-imidazole, 1-trifluoroacetyl, unii-8c6041ow3i, tfai, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethanone, n-trifluoroacetyl imidazole, 2,2,2-trifluoro-1-imidazol-1-yl ethanone, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethan-1-one | |
| C1=CN(C=N1)C(=O)C(F)(F)F | |
| 164.087 | |
| 164.09 | |
| Liquid |
Chemical Identifiers
| 1546-79-8 | |
| 164.087 | |
| SINBGNJPYWNUQI-UHFFFAOYSA-N | |
| 73767 | |
| C1=CN(C=N1)C(=O)C(F)(F)F |
| C5H3F3N2O | |
| MFCD00014501 | |
| 1-trifluoroacetyl imidazole, n-trifluoroacetylimidazole, 1-trifluoroacetyl-1h-imidazole, 1h-imidazole, 1-trifluoroacetyl, unii-8c6041ow3i, tfai, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethanone, n-trifluoroacetyl imidazole, 2,2,2-trifluoro-1-imidazol-1-yl ethanone, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethan-1-one | |
| 2,2,2-trifluoro-1-imidazol-1-ylethanone |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator