missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoroacetyl)imidazole 98.0+%, TCI America™

Catalog No. T06705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06705G 5 g
T067025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T06705G Supplier TCI America Supplier No. T06705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1546-79-8
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.087
MDL Number MFCD00014501
InChI Key SINBGNJPYWNUQI-UHFFFAOYSA-N
Synonym 1-trifluoroacetyl imidazole, n-trifluoroacetylimidazole, 1-trifluoroacetyl-1h-imidazole, 1h-imidazole, 1-trifluoroacetyl, unii-8c6041ow3i, tfai, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethanone, n-trifluoroacetyl imidazole, 2,2,2-trifluoro-1-imidazol-1-yl ethanone, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethan-1-one
PubChem CID 73767
IUPAC Name 2,2,2-trifluoro-1-imidazol-1-ylethanone
SMILES C1=CN(C=N1)C(=O)C(F)(F)F

Specifications

Boiling Point 83°C
Quantity 5 g
UN Number 1993
Formula Weight 164.09
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-(Trifluoroacetyl)imidazole
TSCA No
Recommended Storage Refrigerator