missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoroacetyl)imidazole, 98+%

Catalog No. AAL0013509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0013509 10 g
AAL0013518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0013509 Supplier Thermo Scientific Chemicals Supplier No. L0013509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1546-79-8 | C5H3F3N2O | 164.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1546-79-8
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.087
MDL Number MFCD00014501
InChI Key SINBGNJPYWNUQI-UHFFFAOYSA-N
Synonym 1-trifluoroacetyl imidazole, n-trifluoroacetylimidazole, 1-trifluoroacetyl-1h-imidazole, 1h-imidazole, 1-trifluoroacetyl, unii-8c6041ow3i, tfai, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethanone, n-trifluoroacetyl imidazole, 2,2,2-trifluoro-1-imidazol-1-yl ethanone, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethan-1-one
PubChem CID 73767
IUPAC Name 2,2,2-trifluoro-1-imidazol-1-ylethanone
SMILES C1=CN(C=N1)C(=O)C(F)(F)F

Specifications

Density 1.441
Boiling Point 45°C to 46°C (14 mmHg)
Flash Point 45°C (113°F)
Odor Pungent
Refractive Index 1.424
Quantity 10 g
UN Number UN1993
Beilstein 608904
Sensitivity Moisture sensitive; May form a precipitate
Formula Weight 164.09
Percent Purity ≥98%
Chemical Name or Material 1-(Trifluoroacetyl)imidazole
Show More Show Less
Hazard Category H226-H302-H314
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 216-282-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only