missing translation for 'onlineSavingsMsg'
Learn More

1-(trans-Cinnamoyl)imidazole, 98%, Thermo Scientific Chemicals

Catalog No. AAL0044106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0044106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0044106 Supplier Thermo Scientific Chemicals Supplier No. L0044106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1138-15-4 | Molecular Formula: C12H10N2O | Molecular Weight: 198.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1138-15-4
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.225
MDL Number MFCD00005288
InChI Key XVGXMXZUJNAGFZ-VOTSOKGWSA-N
Synonym n-trans-cinnamoylimidazole, trans-cinnamoylimidazole, imidazole, 1-cinnamoyl-, e, imidazole, 1-cinnamoyl-, trans, imidazole, trans, 1-trans-cinnamoyl imidazole, imidazole, e, e-1-1h-imidazol-1-yl-3-phenylprop-2-en-1-one, 1h-imidazole, e, 2e-1-imidazolyl-3-phenylprop-2-en-1-one
PubChem CID 5357650
IUPAC Name (E)-1-imidazol-1-yl-3-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)N2C=CN=C2

Specifications

Melting Point 128°C to 132°C
Odor Odorless
Quantity 5 g
Beilstein 608846
Sensitivity Moisture sensitive
Formula Weight 198.23
Percent Purity 98%
Chemical Name or Material 1-(trans-Cinnamoyl)imidazole
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 214-510-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only