missing translation for 'onlineSavingsMsg'
Learn More

G-Biosciences 1% Toluidine Blue O, Aqueous

Catalog No. S000789
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000789 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000789 Supplier G-Biosciences Supplier No. 7862205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Blue-colored 1% Toluidine Blue O, Aqueous solution is used to effectively visualize cartilaginous- and chondrogenic-differentiated tissues.

It is used for identifying acidic components in tissues like DNA, RNA, and mast cells. This dye exhibits metachromatic properties, which can stain different components with different colors.

Specifications

Chemical Name or Material Toluidine Blue O
CAS 92-31-9
Molecular Weight (g/mol) 305.8 g/mol
Percent Purity >80%
Grade Research
Color Blue
pH <4.0
Quantity 250 mL