missing translation for 'onlineSavingsMsg'
Learn More

1-(Thien-2-ylmethyl)piperidine-4-carboxylic acid hydrochloridehydrate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 944450-84-4
Molecular Formula C11H18ClNO3S
Molecular Weight (g/mol) 279.779
MDL Number MFCD09817555
InChI Key YUUKLZNSOZECHH-UHFFFAOYSA-N
Synonym 1-thien-2-ylmethyl piperidine-4-carboxylic acid hydrochloride hydrate, 1-thiophen-2-ylmethyl piperidine-4-carboxylic acid hydrate hydrochloride, 1-thiophen-2-ylmethyl piperidine-4-carboxylic acid hydrochloride hydrate, 1-thiophen-2-yl methyl piperidine-4-carboxylic acid-hydrogen chloride-water 1/1/1
PubChem CID 44118590
IUPAC Name 1-(thiophen-2-ylmethyl)piperidine-4-carboxylic acid;hydrate;hydrochloride
SMILES C1CN(CCC1C(=O)O)CC2=CC=CS2.O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC66751CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC66751DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

944450-84-4
279.779
YUUKLZNSOZECHH-UHFFFAOYSA-N
44118590
C1CN(CCC1C(=O)O)CC2=CC=CS2.O.Cl
C11H18ClNO3S
MFCD09817555
1-thien-2-ylmethyl piperidine-4-carboxylic acid hydrochloride hydrate, 1-thiophen-2-ylmethyl piperidine-4-carboxylic acid hydrate hydrochloride, 1-thiophen-2-ylmethyl piperidine-4-carboxylic acid hydrochloride hydrate, 1-thiophen-2-yl methyl piperidine-4-carboxylic acid-hydrogen chloride-water 1/1/1
1-(thiophen-2-ylmethyl)piperidine-4-carboxylic acid;hydrate;hydrochloride

Specifications

944450-84-4
100.0
C11H18ClNO3S
1-thien-2-ylmethyl piperidine-4-carboxylic acid hydrochloride hydrate, 1-thiophen-2-ylmethyl piperidine-4-carboxylic acid hydrate hydrochloride, 1-thiophen-2-ylmethyl piperidine-4-carboxylic acid hydrochloride hydrate, 1-thiophen-2-yl methyl piperidine-4-carboxylic acid-hydrogen chloride-water 1/1/1
C1CN(CCC1C(=O)O)CC2=CC=CS2.O.Cl
279.779
279.79
1-(Thien-2-ylmethyl)piperidine-4-carboxylic acid hydrochloridehydrate
96.5
97%
MFCD09817555
YUUKLZNSOZECHH-UHFFFAOYSA-N
1-(thiophen-2-ylmethyl)piperidine-4-carboxylic acid;hydrate;hydrochloride
44118590
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents