missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecyne, tech. 90%

Catalog No. AAL0257306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0257306 5 g
AAL0257314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0257306 Supplier Thermo Scientific Chemicals Supplier No. L0257306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 765-10-6 | C14H26 | 194.362 g/mol

1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

Solubility
Not miscible with water. soluble in common organic solvents of low polarity

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 765-10-6
Molecular Formula C14H26
Molecular Weight (g/mol) 194.362
MDL Number MFCD00015076
InChI Key DZEFNRWGWQDGTR-UHFFFAOYSA-N
Synonym 1-tetradecyne, tetradecyne, tetradec-1-in, acmc-1bfxn
PubChem CID 69823
IUPAC Name tetradec-1-yne
SMILES CCCCCCCCCCCCC#C

Specifications

Density 0.79
Boiling Point 132°C to 134°C (14 mmHg)
Refractive Index 1.441
Quantity 5 g
Beilstein 1746546
Solubility Information Not miscible with water. soluble in common organic solvents of low polarity
Formula Weight 194.36
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Tetradecyne
EINECSNumber 212-139-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only