missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Tetradecyne 95.0+%, TCI America™
Supplier: TCI America T076125ML
Specifications
| 1-Tetradecyne | |
| 0°C | |
| 252°C | |
| MFCD00015076 | |
| 1-tetradecyne, tetradecyne, tetradec-1-in, acmc-1bfxn | |
| CCCCCCCCCCCCC#C | |
| 194.362 | |
| 194.36 | |
| Liquid |
| 765-10-6 | |
| Yellow | |
| C14H26 | |
| 25 mL | |
| DZEFNRWGWQDGTR-UHFFFAOYSA-N | |
| tetradec-1-yne | |
| 69823 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 765-10-6 | |
| 194.362 | |
| DZEFNRWGWQDGTR-UHFFFAOYSA-N | |
| 69823 | |
| CCCCCCCCCCCCC#C |
| C14H26 | |
| MFCD00015076 | |
| 1-tetradecyne, tetradecyne, tetradec-1-in, acmc-1bfxn | |
| tetradec-1-yne |
Safety and Handling
TSCA : Yes