missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecyne 95.0+%, TCI America™

Catalog No. T07615ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07615ML 5 mL
T076125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07615ML Supplier TCI America Supplier No. T07615ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 765-10-6
Molecular Formula C14H26
Molecular Weight (g/mol) 194.362
MDL Number MFCD00015076
InChI Key DZEFNRWGWQDGTR-UHFFFAOYSA-N
Synonym 1-tetradecyne, tetradecyne, tetradec-1-in, acmc-1bfxn
PubChem CID 69823
IUPAC Name tetradec-1-yne
SMILES CCCCCCCCCCCCC#C

Specifications

Melting Point 0°C
Color Yellow
Boiling Point 252°C
Quantity 5 mL
Formula Weight 194.36
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Tetradecyne