missing translation for 'onlineSavingsMsg'
Learn More

(1-Tetradecyl)trimethylammonium chloride, 98%

Catalog No. AAL1248414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1248414 25 g
AAL1248406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1248414 Supplier Thermo Scientific Chemicals Supplier No. L1248414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4574-04-3 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4574-04-3
MDL Number MFCD00059968

Specifications

Molecular Formula C6H10O2
Quantity 25 g
Beilstein 3916572
Sensitivity Hygroscopic
Synonym cyclopropanecarboxylic acid ethyl ester, ethyl cyclopropylcarboxylate, ethyl cyclopropane carboxylate, cyclopropanecarboxylic acid, ethyl ester, pubchem11840, ethyl cylopropylcarboxylate, ethoxycarbonyl cyclopropane, ethylcyclopropane carboxylate, ksc497s5p, lddosdvzpsglfz-uhfffaoysa
InChI Key LDDOSDVZPSGLFZ-UHFFFAOYSA-N
SMILES CCOC(=O)C1CC1
IUPAC Name ethyl cyclopropanecarboxylate
Molecular Weight (g/mol) 114.14
PubChem CID 78351
Formula Weight 291.95
Percent Purity 98%
Chemical Name or Material (1-Tetradecyl)trimethylammonium chloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-958-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only