missing translation for 'onlineSavingsMsg'
Learn More

(1-Tetradecyl)trimethylammonium chloride, 98%

Catalog No. AAL1248406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1248406 5 g
AAL1248414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1248406 Supplier Thermo Scientific Chemicals Supplier No. L1248406

CAS: 4574-04-3 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4574-04-3
MDL Number MFCD00059968

Specifications

Molecular Formula C17H38ClN
Quantity 5 g
Beilstein 3916572
Sensitivity Hygroscopic
Synonym tetradecyltrimethylammonium chloride, myristyltrimethylammonium chloride, n,n,n-trimethyltetradecan-1-aminium chloride, trimethyltetradecylammonium chloride, 1-tetradecanaminium, n,n,n-trimethyl-, chloride, n-tetradecyltrimethylammonium chloride, rmv7nju3fx, unii-rmv7nju3fx, trimethyl tetradecyl azanium chloride, tetradecyl trimethyl ammonium chloride
InChI Key CEYYIKYYFSTQRU-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCC[N+](C)(C)C.[Cl-]
IUPAC Name trimethyl(tetradecyl)azanium;chloride
Molecular Weight (g/mol) 291.95
PubChem CID 20708
Formula Weight 291.95
Percent Purity 98%
Chemical Name or Material (1-Tetradecyl)trimethylammonium chloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-958-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only