missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecene 90.0+%, TCI America™

Catalog No. T008725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T008725ML 25 mL
T0087500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T008725ML Supplier TCI America Supplier No. T008725ML

Chemical Identifiers

CAS 1120-36-1
Molecular Formula C14H28
Molecular Weight (g/mol) 196.378
MDL Number MFCD00008981
InChI Key HFDVRLIODXPAHB-UHFFFAOYSA-N
Synonym 1-tetradecene, n-tetradec-1-ene, tetradecene, 1-tetradecylene, alpha-tetradecene, unii-fw23481s7s, ccris 3785, alkenes, c10-16 .alpha., a-tetradecene, tetradecene-1
PubChem CID 14260
ChEBI CHEBI:77505
IUPAC Name tetradec-1-ene
SMILES CCCCCCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 232°C
Quantity 25 mL
Formula Weight 196.38
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Tetradecene
EINECSNumber (2)-0027
TSCA Yes