missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecanol, 97+%

CAS: 112-72-1 | C14H30O | 214.39 g/mol

Supplier:  Thermo Scientific Chemicals A1963836

 View more versions of this product

Catalog No. AAA1963836


Only null left
Explore available promotions

Description

Description

1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.

Solubility
Insoluble in water. Soluble in diethyl ether. Slightly soluble in ethanol.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides. Store in a cool place.
Specifications

Specifications

1-Tetradecanol
37°C to 40°C
289°C
C14H30O
500 g
14,6334
Insoluble in water. Soluble in diethyl ether. Slightly soluble in ethanol.
CCCCCCCCCCCCCCO
214.39
CHEBI:77417
≥97%
112-72-1
0.823
141°C (285°F)
MFCD00004757
1742652
1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks
HLZKNKRTKFSKGZ-UHFFFAOYSA-N
tetradecan-1-ol
8209
214.39

Chemical Identifiers

112-72-1
214.39
HLZKNKRTKFSKGZ-UHFFFAOYSA-N
8209
tetradecan-1-ol
C14H30O
MFCD00004757
1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks
CHEBI:77417
CCCCCCCCCCCCCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 204-000-3

RTECSNumber : XB8655000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only