missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecanol, 97+%

Catalog No. AAA1963822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1963822 100 g
AAA1963836 500 g
AAA196380E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1963822 Supplier Thermo Scientific Chemicals Supplier No. A1963822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-72-1 | C14H30O | 214.39 g/mol

1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.

Solubility
Insoluble in water. Soluble in diethyl ether. Slightly soluble in ethanol.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides. Store in a cool place.

Chemical Identifiers

CAS 112-72-1
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.39
MDL Number MFCD00004757
InChI Key HLZKNKRTKFSKGZ-UHFFFAOYSA-N
Synonym 1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks
PubChem CID 8209
ChEBI CHEBI:77417
IUPAC Name tetradecan-1-ol
SMILES CCCCCCCCCCCCCCO

Specifications

Melting Point 37°C to 40°C
Density 0.823
Boiling Point 289°C
Flash Point 141°C (285°F)
Quantity 100 g
Beilstein 1742652
Merck Index 14,6334
Solubility Information Insoluble in water. Soluble in diethyl ether. Slightly soluble in ethanol.
Formula Weight 214.39
Percent Purity ≥97%
Chemical Name or Material 1-Tetradecanol
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 204-000-3
RTECSNumber XB8655000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only