missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecanethiol, 94%

Catalog No. p-7045099
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0940106 5 g
AAL0940114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0940106 Supplier Thermo Scientific Chemicals Supplier No. L0940106
Only null left

CAS: 2079-95-0 | C14H30S | 230.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2079-95-0
Molecular Formula C14H30S
Molecular Weight (g/mol) 230.454
MDL Number MFCD00022099
InChI Key GEKDEMKPCKTKEC-UHFFFAOYSA-N
Synonym 1-tetradecanethiol, myristyl mercaptan, tetradecanethiol, n-tetradecyl mercaptan, tetradecyl mercaptan, 1-mercaptotetradecane, unii-apy3p7s1xm, apy3p7s1xm, tetradecanethiol-1, acmc-1cq2y
PubChem CID 64677
IUPAC Name tetradecane-1-thiol
SMILES CCCCCCCCCCCCCCS

Specifications

Melting Point 6°C to 7°C
Density 0.85
Boiling Point 176°C to 180°C (22 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.461
Linear Formula CH3(CH2)13SH
Quantity 5 g
Beilstein 1742653
Sensitivity Air Sensitive
Formula Weight 230.46
Percent Purity 94%
Chemical Name or Material 1-Tetradecanethiol
Show More Show Less
EINECSNumber 218-209-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only