missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Tetracosanol 98.0+%, TCI America™
$419.69 - $1459.28
Chemical Identifiers
| CAS | 506-51-4 |
|---|---|
| Molecular Formula | C24H50O |
| Molecular Weight (g/mol) | 354.66 |
| MDL Number | MFCD00042662 |
| InChI Key | TYWMIZZBOVGFOV-UHFFFAOYSA-N |
| Synonym | 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc |
| PubChem CID | 10472 |
| ChEBI | CHEBI:77413 |
| IUPAC Name | tetracosan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCO |
Chemical Identifiers
| 506-51-4 | |
| 354.66 | |
| TYWMIZZBOVGFOV-UHFFFAOYSA-N | |
| 10472 | |
| tetracosan-1-ol |
| C24H50O | |
| MFCD00042662 | |
| 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc | |
| CHEBI:77413 | |
| CCCCCCCCCCCCCCCCCCCCCCCCO |
Specifications
| 506-51-4 | |
| White-Yellow | |
| MFCD00042662 | |
| 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc | |
| CCCCCCCCCCCCCCCCCCCCCCCCO | |
| 354.66 | |
| CHEBI:77413 | |
| ≥98.0% (GC) | |
| 1-Tetracosanol |
| 74°C | |
| C24H50O | |
| 1 g | |
| TYWMIZZBOVGFOV-UHFFFAOYSA-N | |
| tetracosan-1-ol | |
| 10472 | |
| 354.66 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes