missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyldimethylsilyl)imidazole 98.0+%, TCI America™

Catalog No. p-7118822
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B10431G 1 g
B10435G 5 g
2 options

Catalog No. B10431G

Supplier: TCI America B10431G

Only null left

Chemical Identifiers

CAS 54925-64-3
Molecular Formula C9H18N2Si
Molecular Weight (g/mol) 182.34
MDL Number MFCD00011682
InChI Key VUENSYJCBOSTCS-UHFFFAOYSA-N
Synonym 1-tert-butyldimethylsilyl-1h-imidazole, 1-tert-butyldimethylsilyl imidazole, tert-butyldimethylsilylimidazole, n-tert-butyldimethylsilylimidazole, t-butyldimethylsilylimidazole, 1-t-butyldimethylsilyl imidazole, 1h-imidazole, 1-1,1-dimethylethyl dimethylsilyl, tbdmsim, imidazole, tbdms derivative, 1-tert-butyldimethylsilylimidazole
PubChem CID 171385
IUPAC Name 1-(tert-butyldimethylsilyl)-1H-imidazole
SMILES CC(C)(C)[Si](C)(C)N1C=CN=C1

Specifications

Color Colorless
Boiling Point 155°C
Quantity 1 g
Formula Weight 182.34
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-(tert-Butyldimethylsilyl)imidazole [tert-Butyldimethylsilylating Agent]