missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-tert-Butyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™
$87.60 - $171.49
Chemical Identifiers
| CAS | 5891-45-2 |
|---|---|
| Molecular Formula | C17H23NO6 |
| Molecular Weight (g/mol) | 337.37 |
| MDL Number | MFCD00077016 |
| InChI Key | VJECGKAFPHEJQS-UHFFFAOYNA-N |
| Synonym | z-glu-otbu, a-tert.butyl ester, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18088, 1-tert-butyl n-cbz-l-glutamate, 1-tert-butyl n-cbz-l-glutamate, z-l-glutamic acid, z-l-glutamic acid |
| PubChem CID | 6994036 |
| IUPAC Name | 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid |
| SMILES | CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 5891-45-2 | |
| 337.37 | |
| VJECGKAFPHEJQS-UHFFFAOYNA-N | |
| 6994036 | |
| CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 |
| C17H23NO6 | |
| MFCD00077016 | |
| z-glu-otbu, a-tert.butyl ester, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18088, 1-tert-butyl n-cbz-l-glutamate, 1-tert-butyl n-cbz-l-glutamate, z-l-glutamic acid, z-l-glutamic acid | |
| 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid |
Specifications
| 5891-45-2 | |
| White | |
| MFCD00077016 | |
| z-glu-otbu, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, z-glu-o-tbu, pubchem18088, pubchem18088, pubchem18089, l-glutamic acid, n-phenylmethoxy carbonyl-, 1-1,1-dimethylethyl ester, z-l-glutamic acid, z-l-glutamic acid | |
| CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 | |
| 337.37 | |
| 337.37 | |
| Crystalline Powder |
| 83°C | |
| C17H23NO6 | |
| 1 g | |
| VJECGKAFPHEJQS-UHFFFAOYNA-N | |
| 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid | |
| 6994036 | |
| ≥98.0% (HPLC,T) | |
| 1-tert-Butyl N-Carbobenzoxy-L-glutamate |
Safety and Handling
TSCA : No