missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-tert-Butoxycarbonylpyrrolidine 98.0+%, TCI America™
$117.44 - $367.70
Chemical Identifiers
| CAS | 86953-79-9 |
|---|---|
| Molecular Formula | C9H17NO2 |
| Molecular Weight (g/mol) | 171.24 |
| MDL Number | MFCD00216581 |
| InChI Key | LPQZERIRKRYGGM-UHFFFAOYSA-N |
| Synonym | 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidine |
| PubChem CID | 643455 |
| IUPAC Name | tert-butyl pyrrolidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCCC1 |
Chemical Identifiers
| 86953-79-9 | |
| 171.24 | |
| LPQZERIRKRYGGM-UHFFFAOYSA-N | |
| 643455 | |
| CC(C)(C)OC(=O)N1CCCC1 |
| C9H17NO2 | |
| MFCD00216581 | |
| 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidine | |
| tert-butyl pyrrolidine-1-carboxylate |
Specifications
| 86953-79-9 | |
| 75°C | |
| MFCD00216581 | |
| 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidine | |
| CC(C)(C)OC(=O)N1CCCC1 | |
| 171.24 | |
| 171.24 | |
| Liquid |
| Colorless | |
| C9H17NO2 | |
| 5 g | |
| LPQZERIRKRYGGM-UHFFFAOYSA-N | |
| tert-butyl pyrrolidine-1-carboxylate | |
| 643455 | |
| ≥98.0% (GC) | |
| 1-tert-Butoxycarbonylpyrrolidine |
Safety and Handling
TSCA : No