missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonylamino)cyclopentanecarboxylic Acid 98.0+%, TCI America™

Catalog No. B38315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38315G 5 g
B38311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38315G Supplier TCI America Supplier No. B38315G
Only null left

Chemical Identifiers

CAS 35264-09-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD01076126
InChI Key YBZCSKVLXBOFSL-UHFFFAOYSA-N
Synonym boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
PubChem CID 2734645
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O

Specifications

Melting Point 133°C
Color White
Quantity 5 g
Formula Weight 229.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonylamino)cyclopentanecarboxylic Acid