missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)piperidine 98.0+%, TCI America™
Supplier: TCI America B462525G
Specifications
| 1-(tert-Butoxycarbonyl)piperidine | |
| Colorless | |
| C10H19NO2 | |
| 25 g | |
| RQCNHUCCQJMSRG-UHFFFAOYSA-N | |
| tert-butyl piperidine-1-carboxylate | |
| 7010304 | |
| ≥98.0% (GC) |
| 75844-69-8 | |
| 120°C | |
| MFCD02093935 | |
| 1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine | |
| CC(C)(C)OC(=O)N1CCCCC1 | |
| 185.267 | |
| 185.27 | |
| Liquid |
Chemical Identifiers
| 75844-69-8 | |
| 185.267 | |
| RQCNHUCCQJMSRG-UHFFFAOYSA-N | |
| 7010304 | |
| CC(C)(C)OC(=O)N1CCCCC1 |
| C10H19NO2 | |
| MFCD02093935 | |
| 1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine | |
| tert-butyl piperidine-1-carboxylate |
Safety and Handling
TSCA : No