missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)piperazine 98.0+%, TCI America™

Catalog No. B24155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24155G 5 g
B241525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24155G Supplier TCI America Supplier No. B24155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57260-71-6
Molecular Formula C9H19N2O2
Molecular Weight (g/mol) 187.26
MDL Number MFCD00075265
InChI Key CWXPZXBSDSIRCS-UHFFFAOYSA-O
Synonym 1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester
PubChem CID 143452
IUPAC Name 4-[(tert-butoxy)carbonyl]piperazin-1-ium
SMILES CC(C)(C)OC(=O)N1CC[NH2+]CC1

Specifications

Melting Point 46°C
Quantity 5 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)piperazine