missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-4-piperidylacetic Acid 97.0+%, TCI America™

Catalog No. B37035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37035G 5 g
B37031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37035G Supplier TCI America Supplier No. B37035G

Chemical Identifiers

CAS 157688-46-5
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD00800239
InChI Key ZXFLMSIMHISJFV-UHFFFAOYSA-N
Synonym 1-boc-4-piperidylacetic acid, 1-boc-piperidin-4-ylacetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl acetic acid, n-boc-4-piperidineacetic acid, 1-boc-4-piperidineacetic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl acetic acid, 1-tert-butoxycarbonyl piperidin-4-yl acetic acid, boc-cmp, boc-cmp-oh
PubChem CID 1512539
IUPAC Name 2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]acetic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)CC(=O)O

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 5 g
Formula Weight 243.30
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-piperidylacetic Acid