missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-4-piperidinecarboxamide 97.0+%, TCI America™

Catalog No. B38921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38921G 1 g
B38925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38921G Supplier TCI America Supplier No. B38921G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91419-48-6
Molecular Formula C11H20N2O3
Molecular Weight (g/mol) 228.292
MDL Number MFCD02180953
InChI Key YHFUWPUJUMZXBD-UHFFFAOYSA-N
Synonym 1-n-boc-piperidine-4-carboxamide, 1-n-boc-isonipecotamide, 1-n-boc-4-piperidinecarboxamide, 1-tert-butoxycarbonyl piperidine-4-carboxamide, n-boc-4-piperidine carboxamide, 1-boc-4-piperidinecarboxamide, 4-carbamoyl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-aminocarbonyl tetrahydropyridine-1 2h-carboxylate, 1-piperidinecarboxylic acid, 4-aminocarbonyl-, 1,1-dimethylethyl ester, tert-butyl 4-carbamoylpiperidinecarboxylate
PubChem CID 2735646
IUPAC Name tert-butyl 4-carbamoylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(=O)N

Specifications

Melting Point 166°C
Color White
Quantity 1 g
Formula Weight 228.29
Percent Purity ≥97.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-piperidinecarboxamide