missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butoxycarbonyl-4-(methylamino)piperidine 98.0+%, TCI America™

Catalog No. B40151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40151G 1 g
B40155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40151G Supplier TCI America Supplier No. B40151G
Only null left

Chemical Identifiers

CAS 147539-41-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD02259411
InChI Key CZYUGTLMFHDODF-UHFFFAOYSA-N
Synonym 1-boc-4-methylaminopiperidine, tert-butyl 4-methylamino piperidine-1-carboxylate, 1-boc-4-methylamino piperidine, 1-boc-4-methylaminopieridine, n-boc-4-methylaminopieridine, n-boc-4-methylaminopiperidine, 4-methylamino-piperidine-1-carboxylic acid tert-butyl ester, 4-methylamino piperidine, n1-boc protected, n-boc-4-n-methylamino piperidine, 4-n-methylamino-1-boc-piperidine
PubChem CID 15380702
IUPAC Name tert-butyl 4-(methylamino)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)NC

Specifications

Color Colorless
Quantity 1 g
Formula Weight 214.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-tert-Butoxycarbonyl-4-(methylamino)piperidine
TSCA No
Recommended Storage Refrigerator