missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-4-hydroxypiperidine 97.0+%, TCI America™

Catalog No. p-7119709
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B26715G 5 g
B267125G 25 g
2 options

Catalog No. B26715G

Supplier: TCI America B26715G

Only null left

Chemical Identifiers

CAS 109384-19-2
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
InChI Key PWQLFIKTGRINFF-UHFFFAOYSA-N
Synonym n-boc-4-hydroxypiperidine, 1-boc-4-hydroxypiperidine, n-boc-4-piperidinol, 1-boc-4-piperidinol, n-boc-4-hydroxylpiperidine, n-boc-4-hydroxy piperidine, 1-tert-butoxycarbonyl-4-hydroxypiperidine, 4-hydroxy-n-boc-piperidine, 1-boc-4-hydroxy-piperidine
PubChem CID 643502
IUPAC Name tert-butyl 4-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)O

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-hydroxypiperidine