missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-pyrrolidinol 98.0+%, TCI America™

Catalog No. B36035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36035G 5 g
B360325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36035G Supplier TCI America Supplier No. B36035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103057-44-9
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD04038535
InChI Key APCBTRDHCDOPNY-UHFFFAOYSA-N
Synonym 1-boc-3-hydroxypyrrolidine, 1-boc-3-pyrrolidinol, n-boc-3-hydroxypyrrolidine, n-boc-3-pyrrolidinol, 1-n-boc-3-hydroxypyrrolidine, 1-pyrrolidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-pyrrolidinol, 3-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-hydroxypyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-hydroxypyrrolidine
PubChem CID 4416939
IUPAC Name tert-butyl 3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)O

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 5 g
Formula Weight 187.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-pyrrolidinol
TSCA No
Recommended Storage Refrigerator