missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-pyrrolidinemethanol 95.0+%, TCI America™

Catalog No. B42495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42495G 5 g
B42491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42495G Supplier TCI America Supplier No. B42495G

Chemical Identifiers

CAS 114214-69-6
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD02179040
InChI Key HKIGXXRMJFUUKV-UHFFFAOYSA-N
Synonym tert-butyl 3-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-3-hydroxymethylpyrrolidine, 1-boc-3-hydroxymethyl pyrrolidine, 3-hydroxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 3-hydroxymethyl-1-boc-pyrrolidine, 1-boc-3-pyrrolidinemethanol, n-boc-dl-beta-prolinol, 1-pyrrolidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, tert-butyl 3-hydroxymethyl-1-pyrrolidinecarboxylate, 3-hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 5237175
IUPAC Name tert-butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)CO

Specifications

Color Yellow
Boiling Point 291°C
Quantity 5 g
Formula Weight 201.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-pyrrolidinemethanol