missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic Acid 95.0+%, TCI America™

Catalog No. B38801G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38801G Supplier TCI America Supplier No. B38801G

Chemical Identifiers

CAS 59378-75-5
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD02179181
InChI Key HRMRQBJUFWFQLX-UHFFFAOYSA-N
Synonym 1-boc-pyrrolidine-3-carboxylic acid, 1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, n-boc-pyrrolidine-3-carboxylic acid, 1-n-boc-beta-proline, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester, 3-carboxy-1-boc-pyrrolidine, 1-tert-butoxy carbonyl pyrrolidine-3-carboxylic acid, 1-n-boc-belta-proline, boc-1-pyrrolidine-3-carboxylic acid, 1-boc-3-pyrrolidinecarboxylic acid
PubChem CID 2760529
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)O

Specifications

Melting Point 135°C
Quantity 1 g
Formula Weight 215.25
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic Acid
TSCA No
Recommended Storage Refrigerator