missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-piperidinemethanol 95.0+%, TCI America™

Catalog No. p-7123167
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37875G 5 g
B378725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37875G Supplier TCI America Supplier No. B37875G
Only null left

Chemical Identifiers

CAS 116574-71-1
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.293
MDL Number MFCD03094733
InChI Key OJCLHERKFHHUTB-UHFFFAOYSA-N
Synonym n-boc-piperidine-3-methanol, tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, 1-boc-3-piperidinemethanol, n-boc-3-piperidinemethanol, 3-hydroxymethyl-1-n-boc-piperidine, n-boc-3-hydroxymethylpiperidine, 1-boc-3-hydroxymethylpiperidine, 1-boc-3-hydroxymethyl piperidine, tert-butyl 3-hydroxymethyl tetrahydro-1 2h-pyridinecarboxylate, +/--n-boc-3-piperidine methanol
PubChem CID 2763851
IUPAC Name tert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CO

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.29
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-piperidinemethanol