missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)-3-hydroxyazetidine 96.0+%, TCI America™
$67.11 - $128.62
Chemical Identifiers
| CAS | 141699-55-0 |
|---|---|
| Molecular Formula | C8H15NO3 |
| Molecular Weight (g/mol) | 173.212 |
| MDL Number | MFCD04115305 |
| InChI Key | XRRXRQJQQKMFBC-UHFFFAOYSA-N |
| Synonym | 1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester |
| PubChem CID | 2756801 |
| IUPAC Name | tert-butyl 3-hydroxyazetidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CC(C1)O |
Chemical Identifiers
| 141699-55-0 | |
| 173.212 | |
| XRRXRQJQQKMFBC-UHFFFAOYSA-N | |
| 2756801 | |
| CC(C)(C)OC(=O)N1CC(C1)O |
| C8H15NO3 | |
| MFCD04115305 | |
| 1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester | |
| tert-butyl 3-hydroxyazetidine-1-carboxylate |
Specifications
| 47°C | |
| White-Yellow | |
| MFCD04115305 | |
| XRRXRQJQQKMFBC-UHFFFAOYSA-N | |
| tert-butyl 3-hydroxyazetidine-1-carboxylate | |
| 173.212 | |
| 173.21 | |
| Crystalline Powder |
| 141699-55-0 | |
| C8H15NO3 | |
| 1-n-boc-3-hydroxyazetidine, n-boc-3-hydroxyazetidine, 1-boc-3-hydroxyazetidine, 1-boc-3-hydroxy azetidine, 1-boc-3-azetidinol, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-azetidin-3-ol, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester | |
| CC(C)(C)OC(=O)N1CC(C1)O | |
| 1 g | |
| 2756801 | |
| ≥96.0% (GC) | |
| 1-(tert-Butoxycarbonyl)-3-hydroxyazetidine |
Safety and Handling
TSCA : No